Hydrochloric acid 36.5 - 38.0% ACS, NF, FCC, VWR Chemicals BDH®
Catalog #: CABDH3026P
Supplier: BDH
CAS Number: 7647-01-0
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Poly bottle
- Size:500 ml
- Supplier No.:3026
Specifications
Specification Test Results
- Assay (HCl):36.5-38.0%
- Assay (HCl) Be':22.7 deg Be' min.
- Ammonium (NH4):0.0003% max.
- Appearance:Clear, colorless fuming liquid
- Arsenic (As):10 ppb max.
- Bromide or Iodide:To pass test
- Clarity of Solution:Free from suspended matter/sediment
- Color (APHA):10 max.
- Free Bromine or Chlorine:To pass test
- Free Chlorine (Cl2):0.0001% max.
- Heavy Metals (as Pb):0.0001% max.
- Identification FCC:To pass test
- Identification NF:To pass test
- Iron (Fe):0.00002% max
- Lead (Pb) FCC/USP:1 mg/kg max.
- Mercury FCC:0.10 ppm max.
- Organic Compounds - Benzene:0.05 ppm max.
- Organic Compounds - Total fluorinated organic compounds:25 ppm max.
- Reducing Substances:To pass test
- Residue After Ignition:0.0005% max.
- Specific Gravity at 60°F:Record only (approx. 1.17-1.20)
- Sulfate (SO4):0.0001% max.
- Sulfite (SO3):0.0001% max.
- Total Organic Compounds:5 ppm max.
- VWR Chemicals BDH®, Hydrochloric acid 36.5 - 38.0% ACS, NF, FCC:
Specification Test Results
About this item
Synonyms: Muriatic acid
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–30 °C
Density:1.18 g/cm³ (20 °C)
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt: 110 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –30 °C
:Flash Pt:
:Flash Pt:
:M.W. 36.46 g/mol
:36.46
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
:8
:II
:GHS05
:GHS07
Customers Who Bought This Also Bought