Order Entry
ContactUsLinkComponent
ANGIOCATH. Fluoropolymer resin I.V. Catheter Placement Unit
BD Angiocath™ IV Catheters, BD™
Catalog #: CABD381123
Supplier:  BD
BD Angiocath™ IV Catheters, BD™
Catalog #: CABD381123
Supplier:  BD
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Length:
    25 mm (1”)
  • Color:
    Blue
  • Cat. no.:
    CABD381123
  • Gauge:
    22
  • Supplier No.:
    381123

Specifications

About this item

These peripheral venous catheters are made of FEP polymer with high lubricity for easy insertion

Tapered catheter tips facilitate penetration and insertion. Instaflash™ Needle Technology provides immediate visual confirmation of vessel entry. Gauge sizes are color-coded for easy identification

Customers Who Bought This Also Bought