Order Entry
ContactUsLinkComponent
Tube Bd Vacutainer Rst 13X100 5.0Ml
Vacutainer® Rapid Serum Tube with Hemogard™ Closure, BD Medical
Catalog #: CABD368774
Supplier:  BD
Tube Bd Vacutainer Rst 13X100 5.0Ml
Vacutainer® Rapid Serum Tube with Hemogard™ Closure, BD Medical
Catalog #: CABD368774
Supplier:  BD
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Ø:
    13 mm
  • Length:
    100 mm
  • Volume:
    5 ml
  • Description:
    Vacutainer® tube
  • Supplier No.:
    368774

Specifications

About this item

BD Vacutainer® Rapid Serum Tubes (RST) contain thrombin-based clot activator and polymer gel for serum separation.

Samples processed in these tubes are used for serum determinations in chemistry. A five-minute clotting time makes this tube ideal for STAT testing in the emergency department as well as clinical laboratories striving to improve test turnaround and workflow efficiencies. This sterile, disposable plastic rapid serum tubes have paper label with orange hemogard closure to use for blood collection. Latex free.

Customers Who Bought This Also Bought