Specifications
- Description:BD Luer-Lok Tip
- Sterility:Sterile
- Cat. no.:CABD309570
- Packaging:100/Box, 800/Case
- Needle Length:5/8"
- Gauge:25
- Supplier No.:309570
Specifications
About this item
Sterile syringe/needle combinations have Slip tip or BD Luer-Lok™ tip.
Thin wall needles have a bigger I.D. without any increase in the O.D. Thin wall needles allow the same flow volume as standard-walled needles one gauge larger.
Sterile syringe/needle combinations have Slip tip or BD Luer-Lok™ tip.
Thin wall needles have a bigger I.D. without any increase in the O.D. Thin wall needles allow the same flow volume as standard-walled needles one gauge larger.
Thin wall needles have a bigger I.D. without any increase in the O.D. Thin wall needles allow the same flow volume as standard-walled needles one gauge larger.
Needles are made of stainless steel with translucent, color-coded polypropylene hubs.