ContactUsLinkComponent
BD SafetyGlide* Specialty Hypodermic Intramuscular Needle
BD SafetyGlide™ Syringes for Insulin, TB, and Allergy, BD
Catalog #: CABD305915
Supplier:  BD
BD SafetyGlide* Specialty Hypodermic Intramuscular Needle
BD SafetyGlide™ Syringes for Insulin, TB, and Allergy, BD
Catalog #: CABD305915
Supplier:  BD
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Color:
    Green Hub
  • Description:
    Needle, Regular Bevel
  • Cat. no.:
    CABD305915
  • Needle Length:
    1"
  • Gauge:
    21
  • Supplier No.:
    305915
Specifications
About this item
The BD SafetyGlide™ Syringe with Tiny Needle Technology completes the BD SafetyGlide line to meet your Sub-Q and intradermal injection needs
  • BD PrecisionGlide™ needle technology
  • One-handed activation
  • Proven dependability
  • Designed for efficiency
SafetyGlide syringes are designed with a permanently attached needle for minimal deadspace and minimal drug waste. The attached needle with Tiny Needle Technology helps meet Sub-Q and intradermal injection needs of the healthcare workers who administer these types of injections. A rotating safety arm allows for low angle injections.
SafetyGlide needles are designed for patient comfort, featuring one-handed activation. They require virtually no change in technique and require minimal training. Finger stays behind the needle at all times. There is virtually no splatter upon activation. Needles are detachable, allowing clinicians to change needles for aspiration and administration.
The permanently attached needle is designed with low waste space which may result in reduced medication waste.
Customers Who Bought This Also Bought