Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Blunt Fill Needle
- Length:38.1 mm (1.50")
- Cat. no.:CABD305180
- Gauge:18
- Supplier No.:305180
Specifications
About this item
Blunt needles require a higher puncture penetration force than a conventional sharp, so they are less likely to cause needlestick injuries while preparing and filling medications from vials, ampoules, and IV bags.
These 18-gauge needles allow for rapid transfer and aspiration without coring and are long enough to easily penetrate single and double-septum barriers. Needles feature a standard Luer hub for standardization and are available already attached to syringes for ready use in medication preparation. The red hub and shield are easily identified as a unique needle.