INLET CO-SEAL,NW40
Catalog #: CAB271-58-453
Supplier: Edwards
Customers Who Bought This Also Bought
Masterflex® L/S® Precision Pump Tubing, FDA-Compliant Viton®, Avantor®
Supplier: VWR International
$268.60
Sodium sulfate, anhydrous ≥99.0%, white powder ACS, VWR Chemicals BDH®
Supplier: VWR International
$109.55 to $1,028.16
Rubidium hydroxide 50% (w/w) in aqueous solution , 99.6% (metals basis)
Supplier: Thermo Scientific Chemicals
$212.00
First Aid Central Compress Pressure Bandages, Trauma Pads, and Non-Adherent Pads, Acme United
Supplier: ACME UNITED LIMITED CA
$1.05
Razor Blades: Double Edge Stainless Steel, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$146.53
Sol-Vet® Blood Collection Needles (Veterinary Care), Sol M
Supplier: SOL-MILLENNIUM MEDICAL
$16.24 to $318.45