Acetone ≥99.5% (by GC, corrected for water content) for HPLC, Supelco®
Catalog #: CAAX0115-1
Supplier: MilliporeSigma
CAS Number: 67-64-1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle
- Size:4 L
- Cat. no.:CAAX0115-1
- Supplier No.:AX0115-1
Specifications
Specification Test Results
- Assay (GC, corrected for water content):99.5% min
- Aldehyde (as HCHO):0.002% max
- Appearance:Clear liquid, free from particulates
- Color (APHA):10 max
- Filtered through 0.2 µm filter:To pass test
- Form:Liquid
- Infrared spectrum:Conforms to standard
- Isopropyl alcohol:0.05% max
- Methanol:0.05% max
- Residue after evaporation:5ppm max
- Solubility in water:To pass test
- Substances reducing permanganate:To pass test
- Titrable acid:0.0003meq/g max
- Titrable base:0.0006meq/g max
- UV Abs. at 330 nm:1.00AU max
- UV Abs. at 340 nm:0.10AU max
- UV Abs. at 350 nm:0.02AU max
- UV Abs. at 400 nm:0.01AU max
- UV Cut-off:330nm max
- Water (H2O):0.5% max
- Acetone ≥99.5%(by GC, corrected for water content) for HPLC, Supelco®:
Specification Test Results
About this item
Synonyms: 2-Propanone,DMK,Propanone
Boiling Pt:56.2 °C (1013 hPa)
Melting Pt:–95.4 °C
Density:0.792 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
Flash Pt:<–20 °C (closed cup)
:Boiling Pt:
:Boiling Pt: 56.2 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -95.4 °C|Melting Pt: –95.4 °C
:Flash Pt:
:Flash Pt: <–20 °C (closed cup)
:M.W. 58.08 g/mol
Merck Index:13,00067
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
REACH:01-2119471330-49
:3
:II
:<–20 °C
:GHS02
:GHS07
Customers Who Bought This Also Bought