Acetic anhydride ≥99.0%, GR ACS, Supelco®
Catalog #: CAAX0080-6
Supplier: MilliporeSigma
CAS Number: 108-24-7
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, pre-packed
- Size:500 ml
- Supplier No.:AX0080-6
Specifications
Specification Test Results
- Assay (GC):99.0% min
- Chloride (Cl):5ppm max
- Form:Clear liquid
- Heavy metals (as Pb):2ppm max
- Infrared spectrum:Conforms to standard
- Iron (Fe):5ppm max
- Phosphate (PO4):0.001% max
- Residue after evaporation:0.003% max
- Substances reducing permanganate:To pass test
- Sulfate (SO4):5ppm max
- Water (H2O):100ppm max
- Supelco®, Acetic anhydride ≥99.0%, GR ACS:
Specification Test Results
About this item
Synonyms: Acetyl oxide
Boiling Pt:139.9 °C (1013 hPa)
Melting Pt:–73 °C
Density:1.08 g/cm³ (20 °C)
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0
Flash Pt:49 °C
:Boiling Pt:
:Boiling Pt: 139.9 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –73 °C
:Flash Pt:
:Flash Pt: 49 °C
:M.W. 102.09 g/mol
Merck Index:13,00057
Storage Temperature:Ambient
EINECS:203-564-8
UN:1715
REACH:01-2119486470-36
:8
:II
:GHS02
:GHS05
:GHS07
Customers Who Bought This Also Bought