ContactUsLinkComponent
BILE ESCULIN AGAR BBL 500GM
Difco™ Bile Esculin Agar Base, BD
Catalog #: CA90005-158
Supplier:  BD
CAS Number:  
Difco™ Bile Esculin Agar Base, BD
Catalog #: CA90005-158
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Difco™ Bile Esculin Agar Base
  • Size:
    500 g
  • Cat. no.:
    CA90005-158
  • Supplier No.:
    299068
Specifications
About this item
Bile Esculin Agar is used to differentiate enterococci and the Streptococcus bovis gropu from other streptococci.
  • Ferric citrate acts as an indicator of esculin hydrolysis
  • Oxgall inhibits gram-positive bacteria other than enterococci
Enterococci are able to split esculin, but other streptococci can not, making esculin hydrolysis a valuable procedure in the identification of enterococci.
Customers Who Bought This Also Bought