ContactUsLinkComponent
TAT Broth Base
Tryptone Azolectin Tween (TAT) Broth Base, BD Biosciences
Catalog #: CA90005-136
Supplier:  BD
CAS Number:  
Tryptone Azolectin Tween (TAT) Broth Base, BD Biosciences
Catalog #: CA90005-136
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Tryptone Azolectin Tween (TAT) Broth Base
  • Size:
    500 g
  • Supplier No.:
    298410
Specifications
About this item
TAT Broth Base is used for cultivating microorganisms from highly viscous or gelatinous materials.
  • Peptone provides nitrogen, vitamins, amino acids, and carbon
  • Especially adapted to the testing of cosmetics
Soy Lecithin and polysorbate 20 neutralize preservatives in the cosmetics or pharmaceutical products, allowing bacteria to grow.
Customers Who Bought This Also Bought