Difco™ Spirit Blue Agar, BD Life Sciences
Catalog #: CA90004-552
Supplier: BD
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Difco™ Spirit Blue Agar
- Size:200 g
- Cat. no.:CA90004-552
- Supplier No.:295020
Specifications
About this item
Spirit Blue Agar is for use with Lipase Reagent for enumeration and detection of lipolytic microorganisms.
- Recommended medium for testing milk and dairy products
- Peptone provides a source of carbon, nitrogen, vitamins, and minerals
- Agar is the solidifying agent
Spirit blue is the indicator of lipolysis.
Customers Who Bought This Also Bought