BD Hycheck™ Paddles for Monitoring Production Environments, BD Diagnostics
Catalog #: CA90004-102
Supplier: BD
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:HYcheck™ for Disinfection Control
- Cat. no.:CA90004-102
- Media:BD trypticase* soy agar and D/E neutralizing agar
- Supplier No.:290002
Specifications
About this item
BD HYcheck™ is a hygiene contact slide used to assess the microbiological contamination of surfaces or fluids
The double-sided slide features a hinged paddle that bends for easy sampling.
Ordering information: Store at 2 to 15°C (36 to 59°F). Multiple media combinations are available; contact your Avantor sales representative for details.
Customers Who Bought This Also Bought