ContactUsLinkComponent
Tryptic Soy Broth without Dextrose
Bacto™ Tryptic Soy Broth without Dextrose, Life Technologies
Catalog #: CA90004-068
Supplier:  BD
CAS Number:  
Bacto™ Tryptic Soy Broth without Dextrose, Life Technologies
Catalog #: CA90004-068
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Bacto™ Tryptic Soy Broth without Dextrose
  • Size:
    500 g
  • Cat. no.:
    CA90004-068
  • Supplier No.:
    286220
Specifications
About this item
Tryptic Soy Broth is a general-purpose liquid enrichment medium used in qualitative procedures for the sterility test and for the enrichment and cultivation of aerobic microorganisms that are not excessively fastidious.
  • Supports the growth of a wide variety of microorganisms
  • Enzymatic digests of casein and soybean meal provide amino acids and nitrogen
  • Lack of glucose means this mixture is low in carbohydrates
Growth in broth media is indicated by the presence of turbidity, specks, or flocculation in the medium while an uninoculated control remains clear and without turbidity after incubation.
Customers Who Bought This Also Bought