ContactUsLinkComponent
D/E Neutralizing Broth
Difco™ D/E (Dey/Engley) Neutralizing Media, BD Life Sciences
Catalog #: CA90004-038
Supplier:  BD
CAS Number:  
Difco™ D/E (Dey/Engley) Neutralizing Media, BD Life Sciences
Catalog #: CA90004-038
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Difco™ D/E (Dey/Engley) Neutralizing Broth
  • Size:
    500 g
  • Cat. no.:
    CA90004-038
  • Supplier No.:
    281910
Specifications
About this item
D/E Neutralizing Agar and D/E Neutralizing Broth have the ability to neutralize antimicrobial chemicals and are used for environmental sampling for the detection and enumeration of microorganisms present on surfaces of sanitary importance.
  • Peptone provides source of nitrogen
  • Yeast extract is a rich source of B-complex vitamins
  • Dextrose is an energy source
The presence and number of microorganisms is determined by the appearance of colonies on the surface of the agar medium.
D/E Neutralizing Broth is used for environmental sampling where neutralization of the chemical is important to determine its bactericidal or bacteriostatic activity. This medium will neutralize a broad spectrum of antiseptic and disinfectant chemicals, including quaternary ammonium compounds, phenolics, iodine and chlorine preparations, mercurials, formaldehyde, and glutaraldehyde.
Customers Who Bought This Also Bought