ContactUsLinkComponent
MacConkey Agar Base
Difco™ MacConkey Agar Base, BD Diagnostics
Catalog #: CA90004-036
Supplier:  BD
CAS Number:  
MacConkey Agar Base
Difco™ MacConkey Agar Base, BD Diagnostics
Catalog #: CA90004-036
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Packaging:
    Bottle
  • Description:
    MacConkey Agar Base
  • Size:
    500 g
  • Cat. no.:
    CA90004-036
  • Supplier No.:
    281810
Specifications
About this item
MacConkey Agar Base is a solid medium recommended for use in qualitative procedures for the cultivation of gram-negative bacilli.
  • Peptones provide nitrogenous nutrients and amino acids
  • Sodium chloride maintains osmotic equilibrium
  • Crystal violet and bile salts inhibit most gram-positive organisms
MacConkey Agar Base is used with added carbohydrate to differentiate enteric gram-negative bacilli based on fermentation reactions.
Customers Who Bought This Also Bought