ContactUsLinkComponent
Dextrose Tryptone Agar
BBL™ Dextrose Tryptone Agar, BD Biosciences
Catalog #: CA90004-020
Supplier:  BD
CAS Number:  
BBL™ Dextrose Tryptone Agar, BD Biosciences
Catalog #: CA90004-020
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    BBL™ Dextrose Tryptone Agar
  • Size:
    500 g
  • Supplier No.:
    280100
Specifications
About this item
Dextrose Tryptone Agar is used for cultivating thermophilic "flat-sour" microorganisms associated with food spoilage.
  • Peptones provide carbon and nitrogen sources
  • Dextrose is the carbohydrate source
  • Bromcresol purple is the pH indicator
  • Agar is the solidifying agent
Dextrose Tryptone Agar can also be used to isolate other food from spoilage bacteria; mesophilic aerobic spore formers in the genera Bacillus and Sporolactobacillus and thermophilic flat sour spore formers such as B. stearothermophilus.
Customers Who Bought This Also Bought