ContactUsLinkComponent
Baird-Parker Agar Base
Difco™ Baird-Parker Agar Base, BD
Catalog #: CA90003-974
Supplier:  BD
CAS Number:  
Difco™ Baird-Parker Agar Base, BD
Catalog #: CA90003-974
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Difco™ Baird-Parker Agar Base
  • Size:
    500 g
  • Supplier No.:
    276840
Specifications
About this item
Baird-Parker Agar Base is used with EY (Egg Yolk) Tellurite Enrichment in the preparation of Egg-Tellurite-Glycerine-Pyruvate Agar (ETGPA) for selective isolation and enumeration of coagulase-positive staphylococci from food, skin, soil, air, and other materials.
  • Peptone, beef extract, and yeast extract provide essential nutrients
  • Sodium pyruvate stimulates growth of S. aureus
  • Egg yolk aids identification by demonstrating lecithinase activity
  • Glycine and lithium chloride inhibit growth of contaminant organisms
Baird-Parker Agar Base may also be used for identification of staphylococci on the basis of their ability to clear egg yolk.
Customers Who Bought This Also Bought