ContactUsLinkComponent
Mueller Hinton Broth
Difco™ Mueller Hinton Broth, BD Life Sciences
Catalog #: CA90003-966
Supplier:  BD
CAS Number:  
Mueller Hinton Broth
Difco™ Mueller Hinton Broth, BD Life Sciences
Catalog #: CA90003-966
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Mueller Hinton Broth
  • Size:
    500 g
  • Cat. no.:
    CA90003-966
  • Supplier No.:
    275730
Specifications
About this item
Mueller Hinton Broth is a general-purpose medium that may be used in the cultivation of a wide variety of fastidious and nonfastidious microorganisms.
  • Casein and beef extract provide essential nitrogen and nutrients
  • Starch protects against toxic substances that may be present
  • Dextrose produced by hydrolysis is a source of energy
Mueller Hinton Broth, unadjusted, has a formula similar to that of the solid medium, but without agar, for use when fluid medium is preferred. It may be used for the general cultivation of bacteria.
Customers Who Bought This Also Bought