ContactUsLinkComponent
Brilliant Green Sulfa Agar
Difco™ BG Sulfa Agar, BD
Catalog #: CA90003-882
Supplier:  BD
CAS Number:  
Difco™ BG Sulfa Agar, BD
Catalog #: CA90003-882
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Difco™ BG Sulfa Agar
  • Size:
    500 g
  • Cat. no.:
    CA90003-882
  • Supplier No.:
    271710
Specifications
About this item
BG Sulfa Agar is a highly selective medium for the isolation of Salmonella.
  • Peptone and yeast extract provide nitrogen, vitamins, and minerals
  • Lactose and sucrose are the sources of carbohydrates
  • Phenol red acts as a pH indicator
Brilliant green and sodium pyridine are complementary in inhibiting gram-positive bacteria and most gram-negative bacilli other than Salmonella species.
Customers Who Bought This Also Bought