ContactUsLinkComponent
Selenite Cystine Broth
Difco™ Fluid Selenite-Cystine Broth, BD
Catalog #: CA90003-808
Supplier:  BD
CAS Number:  
Difco™ Fluid Selenite-Cystine Broth, BD
Catalog #: CA90003-808
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Difco™ Fluid Selenite-Cystine Medium
  • Size:
    500 g
  • Cat. no.:
    CA90003-808
  • Supplier No.:
    268740
Specifications
About this item
Fluid Selenite-Cystine Broth is used as an enrichment medium for isolation of Salmonella species from foods, dairy products, materials of sanitary importance, and clinical specimens.
  • Selective enrichment for the cultivation of Salmonella species
  • Must not be autoclaved
  • Casein peptone provides essential nitrogenous and carbon compounds
Selenite-Cystine Broth is used as an enrichment medium for the isolation of Salmonella from feces, urine, water, foods, and other materials of sanitary importance.
Customers Who Bought This Also Bought