Difco™ Middlebrook 7H10 Agar, BD
Catalog #: CA90003-728
Supplier: BD
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Middlebrook 7H10 Agar
- Size:500 g
- Cat. no.:CA90003-728
- Supplier No.:262710
Specifications
About this item
Middlebrook 7H10 Agar is used in qualitative procedures for the isolation and cultivation of mycobacteria.
- Inorganic salts provides essential substances
- Sodium citrate holds certain inorganic cations in solution
- Glycerol provides plenty of carbon and energy
Supplementation of the agar base is required in order to obtain mycobacterial growth.
Customers Who Bought This Also Bought