ContactUsLinkComponent
SACHETS CAMPY MEDIUM PK20
BD GasPak™ EZ Container Systems, BD Diagnostics
Catalog #: CA90003-646
Supplier:  BD
SACHETS CAMPY MEDIUM PK20
BD GasPak™ EZ Container Systems, BD Diagnostics
Catalog #: CA90003-646
Supplier:  BD
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Campy Sachets, Microaerophilic, Pack of 20
  • Supplier No.:
    260680
Specifications
About this item
Rapid gas generating sachets produce optimal bacterial growth in anaerobic, microaerophilic (reduced oxygen), or CO2 enriched environments.
Pouch systems include 20 resealable pouches and 20 rapid gas-generating sachets. The Anaerobe Pouch System also includes 20 dry anaerobic indicators. Sachets do not require water or a catalyst to produce optimal bacterial growth in anaerobic, microaerophilic (reduced oxygen), or CO2 enriched environments. Pouches are one-step resealable; no sealing bars needed. Multi-layer plastic pouch material preserves environmental integrity for maximum performance. Pouches can accommodate up to four Petri dishes.
Sachets do not require water or a catalyst. Two incubation container sizes are available: standard and large. These containers are nonbreakable and chemical-resistant for maximum safety. Easy-close latches assure a secure, airtight seal. Containers are designed to hold either petri dish racks or standard test tube racks for greater convenience. Standard container and rack holds 15 to 18 petri dishes and the large container and rack holds 30 to 33 petri dishes.
Ordering information: When using the anaerobic system, an anaerobic indicator must be used. Dry indicator strips change from blue to colorless in the absence of oxygen and may be stored at room temperature prior to use.
Customers Who Bought This Also Bought