ContactUsLinkComponent
Antibiotic Medium 11
Difco™ Antibiotic Medium, BD Life Sciences
Catalog #: CA90003-548
Supplier:  BD
CAS Number:  
Difco™ Antibiotic Medium, BD Life Sciences
Catalog #: CA90003-548
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Difco™ antibiotic medium 9
  • Size:
    500 g
  • Cat. no.:
    CA90003-548
  • Supplier No.:
    259310
Specifications
About this item
Antibiotic assay media are used for determining antibiotic potency by the microbiological assay technique.
  • Potency is tested by demonstrating its inhibitory effect
Antibiotic medium 1 and antibiotic medium 4 are used in a cylinder plate method for detecting penicillin in nonfat dry milk.
Antibiotic medium 12 is prepared with the grove and randall formula. It is used for the assay of amphotericin B. Recommended for use in preparing test plates for the cylinder plate assay of nystatin and anisomycin.
Antibiotic medium 19 corresponds to the use described in outline of details for official microbiological assays of antibiotics
Cylinder plate assays are used for commercial preparations of antibiotics, as well as in the quantitative determination of antibioitics in body fluids, animal feeds, and other materials.
Certifications: Antibiotic media are prepared according to the specifications of the USP, European Pharmacopeia, and AOAC International.
Customers Who Bought This Also Bought