Difco™ Anaerobic Agar, BD Diagnostics
Catalog #: CA90003-474
Supplier: BD
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Difco™ Anaerobic Agar
- Size:500 g
- Cat. no.:CA90003-474
- Supplier No.:253610
Specifications
About this item
Anaerobic Agar is used for cultivating anaerobic microorganisms.
- Peptone provides the nitrogen, vitamins, and amino acids
- Dextrose is a carbon source
- Sodium chloride maintains the osmotic equilibrium
- Sodium thioglycollate and sodium formaldehyde sulfoxylate are reducing agents
- Methylene blue serves as indicator of oxygen production
- Agar is the solidifying agent
Methylene blue serves as an indicator of anaerobiosis with a blue color indicating the presence of oxygen.
Customers Who Bought This Also Bought