Order Entry
ContactUsLinkComponent
Todd Hewitt Broth
Bacto™ Todd Hewitt Broth, BD Diagnostics
Catalog #: CA90003-430
Supplier:  BD
CAS Number:  
Bacto™ Todd Hewitt Broth, BD Diagnostics
Catalog #: CA90003-430
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Bacto™ Todd Hewitt Broth
  • Size:
    500 g
  • Cat. no.:
    CA90003-430
  • Supplier No.:
    249240

Specifications

About this item

Todd Hewitt Broth is a general-purpose medium, which primarily is used for the cultivation of beta-hemolytic strepto-cocci, especially for serological studies.

  • Contains peptones, dextrose, and salts for promoting general growth
  • Dehydrated apperarance: Light beige, free-flowing

Selective enrichment broths include the advantages of both enrichment and selection by providing conditions conducive to the growth of group B streptococci while inhibiting the growth of contaminants.

Customers Who Bought This Also Bought