ContactUsLinkComponent
Brain Heart Infusion
Bacto™ Brain Heart Infusion, BD Diagnostics
Catalog #: CA90003-040
Supplier:  BD
CAS Number:  
Bacto™ Brain Heart Infusion, BD Diagnostics
Catalog #: CA90003-040
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Bacto™ Brain Heart Infusion
  • Size:
    500 g
  • Cat. no.:
    CA90003-040
  • Supplier No.:
    237500
Specifications
About this item
Brain Heart Infusion (BHI) Broth is a general-purpose liquid medium for the growth of a wide variety of bacterial and fungal species.
  • Nutrients come from peptones and infusions of brain and heart
  • Supports the growth of fastidious and nonfastidious microorganisms
BHI Broth is used for the cultivation of a wide variety of microorganisms, including bacteria, yeasts, and molds.
Customers Who Bought This Also Bought