ContactUsLinkComponent
BBL Sensi-Disc Susceptibility Test Discs
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, BD Diagnostics
Catalog #: CA90002-496
Supplier:  BD
CAS Number:  
BBL Sensi-Disc Susceptibility Test Discs
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, BD Diagnostics
Catalog #: CA90002-496
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Cefpodoxime
  • Volume:
    10 µg
  • Supplier No.:
    231674
Specifications
About this item
The 6 mm diameter (¹/₄") discs are made from high quality absorbent paper that has been impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents. Discs are clearly marked on both sides with letters and numbers designating the agent and the drug content.
Ordering information: Discs are packaged 50 discs per cartridge. The last disc in each cartridge is marked with an 'X' and contains the agent as coded. Cartridges are for use in BD BBL™ Sensi-Disc™ dispensers.
Customers Who Bought This Also Bought