Diagnostic Tests—Salmonella H Antiserum b to Vibrio cholerae Antiserum, BD Diagnostics
Catalog #: CA90001-826
Supplier: BD
CAS Number:
Specifications
- Description:Salmonella O antiserum, poly A
- Size:3 ml
- Cat. no.:CA90001-826
- Supplier No.:225341
Specifications
About this item
Antigens and antisera used for slide agglutination tests and immunofluorescence.
Store at 2 to 8 °C (35.6 to 46.4 °F).
Customers Who Bought This Also Bought