Order Entry
ContactUsLinkComponent
MacConkey Broth
Difco™ MacConkey Broth, BD
Catalog #: CA90001-042
Supplier:  BD
CAS Number:  
MacConkey Broth
Difco™ MacConkey Broth, BD
Catalog #: CA90001-042
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    MacConkey Broth
  • Size:
    500 g
  • Cat. no.:
    CA90001-042
  • Supplier No.:
    220100

Specifications

About this item

MacConkey Broth is used for the detection of coliform organisms in milk and water.

  • Peptone provides amino acids and other growth factors
  • Lactose is a carbon energy source
  • Oxgall inhibits the growth of gram-positive organisms
  • Bromcresol purple is the indicator

MacConkey Broth is a modification of the original bile salt broth recommended by MacConkey that contained 0.5% sodium taurocholate and litmus as an indicator.

Customers Who Bought This Also Bought