ContactUsLinkComponent
Rappaport-Vassiliadis R10 Broth
Rappaport-Vassiliadis R10 Broth, BD Diagnostics
Catalog #: CA90001-010
Supplier:  BD
CAS Number:  
Rappaport-Vassiliadis R10 Broth, BD Diagnostics
Catalog #: CA90001-010
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Rappaport-Vassiliadis R10 Broth
  • Size:
    500 g
  • Supplier No.:
    218581
Specifications
About this item
Rappaport-Vassiliadis R10 Broth is a liquid medium for selectively enriching Salmonella from meat and dairy products, feces, and polluted water.
  • Tryptone provides carbon and nitrogen for general growth
  • Malachite green is inhibitory to organisms other than Salmonella
  • Low pH helps select for Salmonella species
Selective media recommended by the European Chocolate Manufacturer's Association and the AOAC for rapid detection of motile Salmonella.
Customers Who Bought This Also Bought