ContactUsLinkComponent
Soluble Starch
Difco™ Soluble Starch, BD Life Sciences
Catalog #: CA90000-952
Supplier:  BD
CAS Number:  
Difco™ Soluble Starch, BD Life Sciences
Catalog #: CA90000-952
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Difco™ Soluble Starch
  • Size:
    500 g
  • Cat. no.:
    CA90000-952
  • Supplier No.:
    217820
Specifications
About this item
Provides starch for hydrolysis, detoxification of metabolic byproducts, and as a carbon source.
  • Can be purposed to a variety of applications
Soluble starch features a high molecular weight and is a water-soluble dextrin that provides a source of starch for microorganisms to convert into energy. It is also useful in iodimetry, as it gives an easily visible purple-black end point in the presence of free iodine.
Customers Who Bought This Also Bought