ContactUsLinkComponent
AGAR MI BOTTLE 500G
Difco™ MI Agar, BD
Catalog #: CA90000-794
Supplier:  BD
CAS Number:  
AGAR MI BOTTLE 500G
Difco™ MI Agar, BD
Catalog #: CA90000-794
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    MI Agar
  • Size:
    500 g
  • Cat. no.:
    CA90000-794
  • Supplier No.:
    214883
Specifications
About this item
MI Agar is a chromogenic/fluorogenic medium used to detect and enumerate Escherichia coli and total coliforms in drinking water by the membrane filtration technique.
  • Peptone provides nitrogen, carbon, and amino acids
  • Yeast extract provides trace elements, vitamins, and amino acids
  • Lactose is a carbon source and fermentable carbohydrate
  • Sodium chloride maintains osmotic equilibrium
MI Agar is approved for use by certified drinking water laboratories for microbial analysis of potable water. Other uses include recreational, surface or marine water, bottled water, groundwater, well water, treatment plant effluents, water from drinking water distribution line, drinking water source water and possibly foods.
Customers Who Bought This Also Bought