ContactUsLinkComponent
BBL Dehydrated Culture Media
BBL™ MacConkey II Agar, BD
Catalog #: CA90000-594
Supplier:  BD
CAS Number:  
BBL Dehydrated Culture Media
BBL™ MacConkey II Agar, BD
Catalog #: CA90000-594
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    MacConkey II agar
  • Size:
    500 g
  • Cat. no.:
    CA90000-594
  • Supplier No.:
    212306
Specifications
About this item
MacConkey II Agar is a selective and differential medium for the detection of coliform organisms and enteric pathogens.
  • Low concentration of bile salts
  • Crystal violet inhibits growth of gram-positive bacteria
  • Lactose and neutral red indicator aid in differentiation of enteric microorganisms
MacConkey II Agar is recommended for use with clinical specimens likely to contain mixed microbial flora, such as urine, because it allows a preliminary gropuing of enteric and other gram-negative bacteria.
Customers Who Bought This Also Bought