ContactUsLinkComponent
Nutrient Agar
Difco™ Nutrient Agar, BD
Catalog #: CA90000-260
Supplier:  BD
CAS Number:  
Nutrient Agar
Difco™ Nutrient Agar, BD
Catalog #: CA90000-260
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Difco™ Nutrient Agar
  • Size:
    2 kg
  • Supplier No.:
    211665
Specifications
About this item
Nutrient Agar is used for the cultivation of bacteria and for the enumeration of organisms in water, sewage, feces, and other materials.
  • Beef extract provides carbohydrates, vitamins, nitrogen, and salts
  • Peptones provide a source of nitrogen
  • Agar is the solidifying agent
Nutrient Agar consists of peptone, beef extract, and agar. This relatively simple formulation provides the nutrients necessary for the replication of a large number of microorganisms that are not excessively fastidious.
Customers Who Bought This Also Bought