Order Entry
ContactUsLinkComponent
Salmonella Shigella Agar
BBL™ Salmonella Shigella Agar (SS Agar), BD
Catalog #: CA90000-240
Supplier:  BD
CAS Number:  
BBL™ Salmonella Shigella Agar (SS Agar), BD
Catalog #: CA90000-240
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    BBL™ Salmonella Shigella Agar (SS Agar)
  • Size:
    500 g
  • Cat. no.:
    CA90000-240
  • Supplier No.:
    211597

Specifications

About this item

Salmonella Shigella Agar is a moderately selective and differential medium for the isolation, cultivation, and differentiation of Gram-negative enteric microorganisms isolated from both clinical and nonclinical specimens.

  • Moderately selective medium
  • Contains bile salts, brilliant green, and citrates

Salmonella Shigella Agar is designated as a moderately selective medium based upon the degree of inhibition of Gram-positive microorganisms which it inhibits due to its content of bile salts, brilliant green, and citrates.

Customers Who Bought This Also Bought