Order Entry
ContactUsLinkComponent
BBL Dehydrated Culture Media
BBL™ Phenylethyl Alcohol Agar, BD Biosciences
Catalog #: CA90000-222
Supplier:  BD
CAS Number:  
BBL™ Phenylethyl Alcohol Agar, BD Biosciences
Catalog #: CA90000-222
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    BBL™ Phenylethyl Alcohol Agar
  • Size:
    500 g
  • Cat. no.:
    CA90000-222
  • Supplier No.:
    211539

Specifications

About this item

Phenylethyl Alcohol Agar with 5% Sheep Blood is used to inhibit gram-negative bacteria, specifically Proteus species, in specimens containing a mixed bacterial flora.

  • Sheep blood enhances the growth of anaerobic bacteria
  • Casein digest and soybean meal provide nutrients
  • Sodium chloride maintains osmotic equilibrium
  • Agar is the solidifying agent

PEA agar is a selective medium that is used for the isolation of gram-positive Staphylococcus and Streptococcus species from clinical specimens or specimens that contain mixtures of bacterial flora.

Customers Who Bought This Also Bought