ContactUsLinkComponent
PHENOL RED BROTH BASE 500GM
BBL™ Phenol Red Broth Base, BD Biosciences
Catalog #: CA90000-208
Supplier:  BD
CAS Number:  
BBL™ Phenol Red Broth Base, BD Biosciences
Catalog #: CA90000-208
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    BBL™ Phenol Red Broth Base
  • Size:
    500 g
  • Cat. no.:
    CA90000-208
  • Supplier No.:
    211506
Specifications
About this item
Phenol Red Broth Base and Phenol Red Broth with carbohydrates are used for the determination of fermentation reactions in the differentiation of microorganisms.
  • Pancreatic digest of casein provides nutrients
  • Phenol red acts as a pH indicator
  • Acid production causes a color change
Phenol Red Broth Base is a complete medium without added carbohydrate. It is used as a negative control for fermentation studies or as a base for the addition of carbohydrates.
Customers Who Bought This Also Bought