ContactUsLinkComponent
BBL Dehydrated Culture Media
BBL™ GN (Gram-Negative) Broth, BD Biosciences
Catalog #: CA90000-138
Supplier:  BD
CAS Number:  
BBL™ GN (Gram-Negative) Broth, BD Biosciences
Catalog #: CA90000-138
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    BBL™ GN (Gram-Negative) Broth
  • Size:
    1 lb
  • Cat. no.:
    CA90000-138
  • Supplier No.:
    211279
Specifications
About this item
Gram-Negative Broth is recommended for the selective enrichment of Shigella and Salmonella in clinical and non-clinical specimens.
  • Selectively enriches Shigella and Salmonella
  • Superior to direct inoculation of media
  • For recovery of Salmonella and Shigella
  • More effective than selenite broth for recovery of Shigella
Casein and meat peptones provide amino acids and other nitrogenous substances to support bacterial growth. Dextrose and mannitol supply the energy source. Gram-positive microorganisms and early multiplication of coliforms are both inhibited by sodium citrate and sodium deoxycholate.
Customers Who Bought This Also Bought