BBL™ Brain Heart Infusion, BD Biosciences
Catalog # CA90000-060
Supplier: BD
CAS Number:
Specifications
- Description:BBL™ brain heart infusion
- Size:500 g
- Cat. no.:CA90000-060
- Supplier No.:211059
Specifications
About this item
Brain Heart Infusion (BHI) broth is a general-purpose liquid medium for the growth of a wide variety of bacterial and fungal species.
- Brain and heart infusions and peptones provide nutrients
- Supports the growth of fastidious and nonfastidious microorganisms
BHI Broth is used for the cultivation of a wide variety of microorganisms, including bacteria, yeasts, and molds.
Brain heart infusion, modified differs from other formulations by the quantities of the ingredients and the substitution of pancreatic digest of casein for pancreatic digest of gelatin.
Frequently Bought Together