ContactUsLinkComponent
Tetrathionate Broth Base
Difco™ Tetrathionate Broth Base, BD Biosciences
Catalog #: CA90000-008
Supplier:  BD
CAS Number:  
Difco™ Tetrathionate Broth Base, BD Biosciences
Catalog #: CA90000-008
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Difco™ Tetrathionate Broth Base
  • Size:
    500 g
  • Cat. no.:
    CA90000-008
  • Supplier No.:
    210430
Specifications
About this item
Tetrathionate Broth Base is used as a selective enrichment medium for the isolation of Salmonella from feces, urine, foods, and other materials of sanitary importance.
  • Peptones provide nitrogen, vitamins, amino acids, and carbon
  • Oxgall inhibits Gram-positive microorganisms
  • Calcium carbonate neutralizes and absorbs toxic metabolites
Tetrathionate, which is formed in the medium by the addition of the iodine-iodide solution, inhibits the normal intestinal flora of fecal specimens.
Customers Who Bought This Also Bought