Order Entry
ContactUsLinkComponent
SPIN FILTER 3K 15ML 24PK
Macrosep® Advance Centrifugal Devices, Cytiva (Formerly Pall Lab)
Catalog #: CA89131-974
Supplier:  Cytiva
SPIN FILTER 3K 15ML 24PK
Macrosep® Advance Centrifugal Devices, Cytiva (Formerly Pall Lab)
Catalog #: CA89131-974
Supplier:  Cytiva
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Capacity:
    20 ml (⅔ oz.)
  • Membrane Material:
    Omega
  • Color:
    Gray
  • Operating temperature range:
    0 to 40°C (32 to 104°F)
  • Overall length:
    12.0 cm (142/3")
  • Effective filtration area:
    7.2 cm2
  • Max. centrifugal force:
    5,000 g (ultrafiltion), 14,000 g (microfiltration)
  • MWCO:
    3K
  • Supplier No.:
    MAP003C37

Specifications

About this item

These centrifugal devices can quickly concentrate up to 20 ml of biological sample without valuable sample loss.

  • Provides high recoveries, typically >90%
  • Low protein-binding Omega™ membrane and polypropylene housing minimize losses due to non-specific binding
  • Versatile ultrafiltration Omega™ membrane is available in a variety of MWCOs
  • Built-in deadstop prevents spinning to dryness
  • Color-coded and laser etched for easy identification

Macrosep™ Advance centrifugal devices can be used for a wide range of ultrafiltration applications including the concentration and desalting of proteins, buffer exchange and protein fractionation.

The design of the devices maximizes the effective filtration area allowing for rapid sample processing while maintaining a gentle concentration environment to preserve protein activity and conformation.

Customers Who Bought This Also Bought