DuPont™ Tyvek® IsoClean® Hoods with Full Face Opening
Catalog #: CA89125-650
Supplier: DuPont
Specifications
- Garment application:Cleanroom
- W×L:281/2×171/2"
- Color:White
- Cat. no.:CA89125-650
- Supplier No.:IC668BWH00CS
Specifications
About this item
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
- Bound seams
- Full face opening
- Bound hood opening
- Ties with loops for adjustable fit
- One size fits most
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration. Bound hood opening ensures lower particle shedding. Full face opening and ties with loops allow for an adjustable fit.
Clean-processed garments are processed to minimize particle shedding.
Certifications: Sterile garments are gamma sterilized to SAL of 10-6.
Packaging: Bulk packaged option packed in double transparent poly liners.
Customers Who Bought This Also Bought