Steraffirm™ Bowie-Dick Test Pack, STERIS®
Catalog #: CA89065-290
Supplier: Steris
Specifications
- Description:Test Packs
- Cat. no.:CA89065-290
- Supplier No.:EQC003
Specifications
About this item
Test packs consist of preassembled single-use test packs designed to evaluate the performance of the air removal system of a pre-vacuum equipped sterilizer.
Note: For use in life sciences applications only. Not approved for use in healthcare facilities in the United States.
Calibrated to be compatible with pre-vacuum cycles operating at specific exposure temperatures or within a specified exposure temperature range
Certifications: Conforms to the EN 867-4: 2001 (Class B) or ANSI/AAMI ST66 standards. No lead or other heavy metals. Abrupt and distinctive total color change from yellow to blue/purple. Can detect "wet steam," superheated steam, and noncondensables in the steam supply.
Customers Who Bought This Also Bought