UltraSense™ EC Powder-Free Nitrile Gloves, Microflex®, Ansell
Catalog #: CA89065-282
Supplier: Microflex
Specifications
- Ambidextrous/Hand-specific:Ambidextrous
- Glove Color:Blue
- Glove size:M
- Length:29 cm (11 3/8")
- Powdered/Powder-free:Powder-free
- Disposable / reusable:Disposable
- Finger thickness:5.9 mil
- Palm thickness:4.7 mil
- Cuff thickness:3.9 mil
- Cat. no.:CA89065-282
- Supplier No.:USE-880-M
Specifications
About this item
Ultra-soft, powder-free gloves are made from a nitrile formulation that has the fit and feel of latex and provides reliable protection against many hazardous and infectious substances.
- Ambidextrous
- Beaded cuffs
- Textured fingers
Non-stick surface resists tapes and adhesives.
Caution: Components used in making these gloves may cause allergic reactions in some users. Follow your institution's policies for use.
Customers Who Bought This Also Bought