KIMBERLY-CLARK® BASIC™ Plus Lab Coats, KIMBERLY-CLARK PROFESSIONAL®
Catalog #: CA89033-030
Supplier: Kimberly-Clark
Specifications
- Clothing Size:XL
- Garment application:Laboratory
- Color:Blue
- Environmentally Preferable:Waste Reducing
- Cat. no.:CA89033-030
- Supplier No.:10033
Specifications
About this item
Lab coats are manufactured from a protective three-layer spunbond-meltblown-spunbond (SMS) fabric for protection against light fluid contact
- Three-layer SMS fabric for protection against light fluid contact
- Five-snap front closure, one breast pocket, two hip pockets, knit collar and cuffs
Lab coats feature a five-snap front closure, one breast pocket, two hip pockets, and knit collar and cuffs.
Customers Who Bought This Also Bought