Jackson Safety® G40 Nitrile Coated Gloves, Kimberly-Clark
Catalog #: CA89003-837
Supplier: Kimberly-Clark
Specifications
- Ambidextrous/Hand-specific:Ambidextrous
- Glove Color:Blue,Black
- Glove size:XL
- Cat. no.:CA89003-837
- Supplier No.:40228
Specifications
About this item
Designed to provide an improved level of performance and quality, the new G40 nitrile coated glove replaces the G40 PURPLE NITRILE™ glove as the optimal choice for jobs with improved strength, grip, and dexterity.
- Gloves ensure a better oil and industrial grease grip
- The washable design maintains the EN388 ratings through at least five wash cycles
- Improved heat transfer stamp, with ratings visible even after glove use
- Ecofriendly packaging with no separate insert, no printed bags, and less waste
- Silicone-free
Typical applications include automotive and aerospace assembly, metal, glass, and wood handling.
Customers Who Bought This Also Bought