To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Dimensions:64L x 76W x 92H mm (21/2 x 3 x 35/8")
- Color:Green
- Description:EasyDip™ Slide Staining Jars
- Cat. no.:CA87000-122
- Supplier No.:M900-12G
Specifications
About this item
The slide staining system is made of acetal plastic.
Jars can be loosely joined to each other laterally to maintain order in the laboratory. Additionally, jars are available in five different colors to assist in identifying contents or applications.
- Jars can be loosely joined to each other to keep the same order when moved around
- Wide stable base for stability while inside is recessed to reduce reagent volume
- Available in five colours to aid identification of contents or applications
The slide staining system is made of acetal plastic.
The staining jar is made of resistant acetal plastic, and will not break like most glass jars do. It will resist most staining agents including alcohol and xylene (but not phenol, iodine, or ferric chloride). The wide stable base offers greater stability while the inside is recessed, allowing for a smaller reagent volume of only 80 ml. Easy to clean and no metals to corrode. Ideal for special stains, frozen sections, and special processes.
The staining jar is made of resistant acetal plastic, and will not break like most glass jars do. It will resist most staining agents including alcohol and xylene (but not phenol, iodine, or ferric chloride). The wide stable base offers greater stability while the inside is recessed, allowing for a smaller reagent volume of only 80 ml. Easy to clean and no metals to corrode. Ideal for special stains, frozen sections, and special processes.
The system is not autoclavable. Volume is 80 ml. System dimensions are 64×76×92 mm (2.5×3×3.625").
The system is not autoclavable. Volume is 80 ml. System dimensions are 64×76×92 mm (2.5×3×3.625").