Face Shields and Visors, 3M™
Catalog #: CA80077-247
Supplier: 3M
Specifications
- Color:Clear
- Description:Polycarbonate faceshield WP98
- Supplier No.:82543-00000
Specifications
About this item
Faceshields provide ventilation and face protection for high heat environments.
- Faceshieldd mount quickly to the appropriate 3M™ headgear without tools, snaps, or cams
- Replaceable eyeshields offer increased visibility
Certifications: Items meet ANSI Z87.1-2003 standards.
Customers Who Bought This Also Bought