ContactUsLinkComponent
Slide Draining Rack
Slide Drying/Draining Holder, JAC Medical Products
Catalog #: CA78010-056
Supplier:  JAC
Slide Drying/Draining Holder, JAC Medical Products
Catalog #: CA78010-056
Supplier:  JAC
Specifications
  • Color:
    Blue
  • Cat. no.:
    CA78010-056
  • L×W×H:
    16.8 x 9.8 x 2.2 cm (65/8x37/8x7/8")
  • Supplier No.:
    JAC22
Specifications
About this item
Ideal for bone marrow smears, this holder accommodates 12 slides individually or 24 slides back-to-back
Holder provides protection from breakage and contamination of slides. Openings in the well bottoms facilitate draining. Constructed of thick, high-quality polypropylene for durability. Autoclavable.
Customers Who Bought This Also Bought